Learn More
Medchemexpress LLC IQ-1S free acid | 23146-22-7 | MFCD00612483 | 99.3% | 247.25 | C15H9N3O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100233100MG
Encompass_Preferred
IQ-1S free acid is a small-molecule research compound that inhibits NF-κB/AP-1 activity and binds c-Jun N-terminal kinases (JNKs). It is used for cellular and biochemical studies of MAPK/JNK and NF-κB/AP-1 signaling. Molecular formula C15H9N3O, CAS 23146-22-7; molecular weight 247.25.
Catalog No. 50-002-15264