Learn More
Medchemexpress LLC IQ-3 10mg | 312538-03-7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY10760010MG
Encompass_Preferred
IQ-3 is a specific inhibitor of the c-Jun N-terminal kinase (JNK) family with preference for JNK3 IQ-3 exhibits Kd values of 0 24 M 0 29 M and 0 066 M for JNK1 JNK2 and JNK3 respectively[1
Catalog No. 50-002-40164