Learn More
Aviva Systems Biology Corp IQCF1 Antibody - middle region (ARP52938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IQCF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IQCF1. Alternate names: FLJ27508, MGC39725ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60873