Learn More
TARGETMOL CHEMICALS INC IQP-0528 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T276255MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IQP-0528 is a potent non-nucleoside reverse transcriptase inhibitor (NNRTI) that has potential for the treatment of HIV infection by blocking viral entry and shows antiviral activity as a microbicidal gel. . IQP-0528 showed inhibition of both HIV-1 and HIV-2 with an EC50 of 0.2 nM for HIV-1 and 100 nM for HIV-2. purity: 98%
Catalog No. 50-258-0596