Learn More
Medchemexpress LLC IR 813 tosylate | 134127-48-3 | MFCD03093252 | 99.5% | 755.41 | C47H47ClN2O3S | 1 ML

Supplier: Medchemexpress LLC HYW24859410MM/1ML
IR 813 tosylate is a near-infrared (NIR) fluorescent dye for in vivo visualization of regional lymph nodes in mice. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO, with supporting analytical and safety documents available.
- Near-infrared fluorescent dye suitable for lymph node imaging in small animals.
- Available as 10 mM in DMSO (1 mL) and as solid in multiple pack sizes.
- High analytical purity (~99.5%).
- Chemical formula C47H47ClN2O3S; molecular weight 755.41 g/mol.
- Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
- Datasheet, certificate of analysis, and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.