Learn More
Strem, An Ascensus Company CAS# 10025-97-5. 250mg. Iridium(IV) chloride, hydrate. MFCD00150243
SDP
Supplier:  Strem, An Ascensus Company 937727250MG
Encompass
CAS# 10025-97-5. 250mg. Iridium(IV) chloride, hydrate. MFCD00150243. Molecular Weight: 334.03. Molecular Formula: IrCl4∙XH2O. Color/form: black pwdr. Strem# 93-7727. http://www.strem.com/catalog/v/93-7727/
Catalog No. 50-204-5429