Learn More
Medchemexpress LLC HY-77048 10mg Medchemexpress, IRAK-4 protein kinase inhibitor 2 CAS:301675-24-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7704810MG
Encompass_Preferred
Medchemexpress, HY-77048 10mg IRAK-4 protein kinase inhibitor 2 CAS:301675-24-1 IRAK-4 protein kinase inhibitor 2 (compound 1) is a potent inhibitor of interleukin-1 (IL-1) receptor-associated kinase-4 (IRAK-4), with an IC50 of 4 μM. IRAK-4 protein kinase inhibitor 2 can be used for the research of inflammatory and immune-related conditions or disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2285