Learn More
TARGETMOL CHEMICALS INC IRAK INHIBITOR 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5094
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IRAK inhibitor 1 is an effective IRAK-4 inhibitor (IC50 216 nM) is poorly active against JNK-1 (IC50 3.801 uM) and JNK-2 (IC50>10 uM). purity: 97%
Catalog No. 50-257-9815