Learn More
Medchemexpress LLC HY-13278A 5mg Medchemexpress, IRAK inhibitor 4 (trans) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13278A5MG
Encompass_Preferred
Medchemexpress, HY-13278A 5mg IRAK inhibitor 4 (trans) CAS: IRAK inhibitor 4 (trans) is the trans form of IRAK inhibitor 4. IRAK inhibitor 4 is an interleukin-1 receptor associated kinase 4 ( IRAK4 ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2658