Learn More
Cayman Chemical IRAK InhIbItr 1 25mg
SDP
Supplier:  Cayman Chemical 3663725 MG
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 35 nM); selective for IRAK4 over JNK1 and JNK2 at 10 µM; prevents increases in TNF-α and IL-12p40 levels in M. bovis-infected THP-1 cells at 1 µM
Catalog No. 50-291-3862