Learn More
Cayman Chemical IRAK14 InhIbItr 5mg
SDP
Supplier:  Cayman Chemical 175405 MG
Encompass_Preferred
A benzimidazole that selectively disrupts the activity of IRAK 1 and 4 (IC50 = 0.3 and 0.2 µM, respectively)
Catalog No. 50-296-5831