Learn More
Medchemexpress LLC HY-101922 5mg Medchemexpress, IRAK4-IN-1 CAS:1820787-94-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1019225MG
Encompass_Preferred
Medchemexpress, HY-101922 5mg IRAK4-IN-1 CAS:1820787-94-7 IRAK4-IN-1 is an interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor with an IC50 of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7917