Learn More
Medchemexpress LLC Irak4-in-14 | 2667681-71-0 | 99.0% | 489.55 g/mol | C25H28FN9O | 5 MG

Supplier: Medchemexpress LLC HY1461125MG
IRAK4-IN-14 is a small-molecule, selective, orally active inhibitor of interleukin-1 receptor-associated kinase 4 (IRAK4) characterized for biochemical potency and in vivo pharmacokinetics. It has been profiled for cellular activity and shows favorable oral exposure in preclinical species.
- Potent IRAK4 inhibition (IC50 0.003 μM).
- Cell pIRAK4 activity (IC50 0.11 μM).
- Selective kinase profile with limited off-target activity reported.
- Oral bioavailability approximately 66% in mouse.
- Molecular formula C25H28FN9O; molecular weight 489.55 g/mol.
- Reported purity 99.01%.
- Available as solid packages (including 5 mg) and as a 10 mM solution in DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.