Learn More
Medchemexpress LLC IRAK4-IN-20 | 1931994-80-7 | C22H25F3N4O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY150735100MG
Encompass_Preferred
IRAK4-IN-20 (Compound BAY-1834845) is an orally active IRAK4 inhibitor with an IC50 of 3.55 nM. It can be used for acute respiratory distress syndrome (ARDS) research. It effectively prevents acute respiratory distress syndrome in mice and decreases inflammatory cytokines secretion effectively in LPS stimulated human peripheral blood mononuclear cells (PBMC).
  • Orally active IRAK4 inhibitor
  • IC50 of 3.55 nM
  • Used for acute respiratory distress syndrome (ARDS) research
  • Decreases inflammatory cytokines secretion in LPS stimulated human PBMC
  • Prevents acute respiratory distress syndrome in mice
Catalog No. 50-003-75498