Learn More
Medchemexpress LLC Irak4-in-21 | 2170694-04-7 | C28H28FN7O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1513631MG
Encompass_Preferred
IRAK4-IN-21 (compound 17) is an orally active, potent, and selective IRAK4 inhibitor. It effectively inhibits IL-23 production and can be used in studies of autoimmune diseases such as plaque psoriasis and psoriatic arthritis.
  • Potent and selective IRAK4 inhibitor with IC50 values of 5 nM for IRAK4 and 56 nM for TAK1.
  • Effectively inhibits IL-23 production (IC50 = 0.17 μM).
  • Decreases levels of IL-23, IL-6, and MIP-1β in various cell lines.
  • Shows moderate efficacy in acute animal model studies for inhibition of IL-6 production.
Catalog No. 50-003-74194