Learn More
Medchemexpress LLC HY-114181 10mg Medchemexpress, IRAK4-IN-4 CAS:1850276-58-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11418110MG
Encompass_Preferred
Medchemexpress, HY-114181 10mg IRAK4-IN-4 CAS:1850276-58-2 IRAK4-IN-4 is an interleukin-1 receptor–associated kinase 4 (IRAK4) inhibitor extracted from patent CN107163044A, Compound15, has an IC50 of 2.8 nM. IRAK4-IN-4 also inhibits cyclic GMP-AMP synthase (cGAS) with an IC50 of 2.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9301