Learn More
Selleck Chemical LLC IRAK4-IN-4 S0775-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S077510MM/1ML
Encompass_Preferred
IRAK4-IN-4 (compound 15) is an inhibitor of interleukin-1 receptor associated kinase 4 (IRAK4) and cyclic GMP-AMP synthase (cGAS) with IC50 of 2 8 nM and 2 1 nM respectively
Catalog No. 50-000-43425