Learn More
Medchemexpress LLC IRAK4-IN-7 | 1801343-74-7 | 99.36% | 417.42 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY10958550MG
Encompass_Preferred
IRAK4-IN-7 is a selective, potent, and orally active interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor, extracted from patent WO2015104688 (example 1). It has potential for cancer and inflammatory disease treatment. It exhibits anti-cancer and anti-inflammatory activities.
  • Selective and potent IRAK4 inhibitor
  • Orally active
  • Potential for cancer treatment
  • Potential for inflammatory disease treatment
  • Exhibits anti-cancer activities
  • Exhibits anti-inflammatory activities
Catalog No. 50-003-44349