Learn More
Medchemexpress LLC IRAK4-IN-7 | 1801343-74-7 | 99.4% | 417.42 | 100 UG
SDP
Supplier:  Medchemexpress LLC HY10958510MG
Encompass_Preferred
IRAK4-IN-7 is a selective, potent, and orally active interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor, extracted from patent WO2015104688 (example 1). It has the potential for cancer and inflammatory diseases treatment. For research use only, not for sale to patients.
  • Selective, potent, and orally active inhibitor.
  • Potential for cancer and inflammatory diseases treatment.
  • Has anti-cancer and anti-inflammatory activities.
Catalog No. 50-003-44728