Learn More
Medchemexpress LLC IRAK4-IN-7 | 1801343-74-7 | 99.4% | 417.42 | 200 UG
SDP
Supplier:  Medchemexpress LLC HY10958510MM/1ML
Encompass_Preferred
IRAK4-IN-7 is a selective, potent, and orally active interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor. It is extracted from patent WO2015104688 (example 1) and holds potential for the treatment of cancer and inflammatory diseases. It exhibits anti-cancer and anti-inflammatory activities, making it suitable for research in these therapeutic areas.
  • Selective and potent inhibitor
  • Orally active
  • Potential for cancer treatment
  • Potential for inflammatory diseases treatment
  • Suitable for in vivo and in vitro research
Catalog No. 50-003-44961