Learn More
Cayman Chemical IRAK4 InhIbItr 1 10mg
SDP
Supplier:  Cayman Chemical 3754810 MG
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 7 nM); reduces resiquimod-induced increases in TNF-α levels in isolated human PBMCs (EC50 = 300 nM)
Catalog No. 50-291-6101