Learn More
CPC Scientific H-Ser-Gly-Ile-Pro-Tyr-Ile-Ile-Ser-Tyr-Leu-His-Pro-Gly-Asn-Thr-Ile-Leu-His-Val-Asp-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific IRBP002B
Encompass
This 20-residue peptide, a major pathogenic T-cell epitope (161?180), is present in the first homologous repeat of the interphotoreceptor retinoid binding protein peptide (IRBP). It has been shown to induce posterior uveitis (EAU).ONE-LETTER SEQUENCE: SGIPYIISYLHPGNTILHVDMOLECULAR FORMULA: C103H157N25O29MOLECULAR WEIGHT:2209.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [211426-18-5], RESEARCH AREA: ImmunomodulationREFERENCES: Sonoda, K. et al. J. Immunol. 171, 2652 (2003); Read, RW. and SR. Barnum, ARVO Meeting Abstracts 43, 2281 (2002); Silver, P. et al. Invest. Ophthal. Vis. Sci. 36, 946 (1995); Karabekian, Z. et al. Invest. Ophthalmol. Vis. Sci. 46, 3769 (2005).
Catalog No. 50-210-9609