Learn More
Cayman Chemical IRBP1-20humnratrI 10mg
SDP
Supplier:  Cayman Chemical 3672710 MG
Encompass_Preferred
A peptide inducer of experimental autoimmune uveoretinitis; increases the levels of IL-4, IL-5, and IL-6 secreted from primary mouse spleen lymphocytes at 3 µM; immunization and rechallenge increases EAr swelling in IFN-γ-deficient mice; induces ocular blood vessel inflammation, immune cell infiltration, and photoreceptor layer damage, as well as retinal detachment, in the same mice
Catalog No. 50-290-9680