Learn More
LICOR IRDye 680LT Donkey anti-Goat IgG Secondary Antibody (0.5 mg)
Supplier:  LICOR 92668024
Encompass_Preferred
Immunogen: Goat IgGPurity and Specificity: Isolation of specific antibodies was accomplished by affinity chromatography using goat IgG covalently linked to agarose. Based on ELISA, this antibody reacts with the heavy and light chains of goat IgG and sheep IgG. This antibody has been tested by dot blot and/or solid-phase adsorbed to ensure minimal cross-reactivity with human, mouse, rabbit, rat, chicken. guinea pig, hamster, swine, and horse serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot applications.
Catalog No. NC0116358