Learn More
LICOR IRDye 680LT Goat anti-Rat IgG Secondary Antibody (0.1 mg)
Supplier:  LICOR 92568029
Encompass_Preferred
Immunogen: Rat IgG, whole molecule | Applications: IRDye 680LT Goat anti-Rat IgG Secondary Antibody is recommended for: Western Blot, Protein Array, Immunohistochemistry, Microscopy, 2D Gel Detection, and Tissue Section Imaging. IRDye 680LT Goat anti-Rat IgG Secondary Antibody is not recommended for Small Animal Imaging, In-Cell Western Assay, and On-Cell Western Assay | Formulation: IRDye 680LT Secondary Antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. NC1536278