Learn More
LICOR IRDye 700 CREB Consensus Oligonucleotide (25 uL)
Supplier:  LICOR 82907923
Encompass_Preferred
IRDye 700 oligonucleotides are supplied as 25 µL of 50 nM (or 50 fmol/µL) double-stranded DNA. CREB proteins are cellular transcription factors activated by phosphorylation of various kinases that bind to certain DNA sequences. For this reason, it is a common oligonucleotide used in EMSA to assess protein binding.
Catalog No. NC2209426