Learn More
LICOR IRDye 800CW Alkyne Infrared Dye (50mg)
Supplier:  LICOR 92965050
Encompass_Preferred
IRDye 800CW Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 800CW.
Catalog No. NC2209472