Learn More
LI-COR IRDye 800CW Carboxylate (100 nmol)
SDP
Supplier:  LI-COR 92908972
Encompass_Preferred
Assays that use IRDye 800CW conjugates (such as in vivo imaging and cell binding assays) may require a "dye-only" control for potential effects or retention of the dye. Carboxylate (non-reactive) form of IRDye 800CW is an ideal control. (Note: The carboxylate dye has no reactive group and cannot be used for labeling).Properties: Chemical Formula: C46H50N2Na4O15S4; Molecular Weight: 1091.10 g/mol; Exact Mass: 1090.17
Catalog No. NC9807392