Learn More
LICOR IRDye 800CW Goat anti-Mouse IgM ( chain specific) Secondary Antibody (0.5 mg)
Supplier:  LICOR 92632280
Encompass_Preferred
Immunogen: Mouse IgM paraproteins | Applications: IRDye 800CW Goat anti-Mouse IgM (µ chain specific) Secondary Antibody is highly recommended for: Western Blot, In-Cell Western™ Assay, On-Cell Western Assay, Protein Array, Immunohistochemistry, Small Animal Imaging, Microscopy, 2D Gel Detection, Tissue Section Imaging, and Virus Titration Assay | Formulation: IRDye 800CW Secondary Antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. 50-489-547