Learn More
Medchemexpress LLC Iretol (2,4,6-trihydroxyanisole) | 487-71-8 | 99.4% | 156.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1393850MG
Encompass_Preferred
Iretol (2,4,6-trihydroxyanisole) is a degradation product of a glucoside obtained from Iris Jorentina. It serves as an intermediate in the synthesis of natural isoflavones.
  • Used to synthesize Tectorigenin, Irigenin, and Caviunin.
  • For research use only.
Catalog No. 50-003-47971