Learn More
Medchemexpress LLC Iretol | 487-71-8 | 99.4% | 156.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13938100MG
Encompass_Preferred
Iretol is a degradation product of a glucoside obtained from Iris Jorentina. It functions as an intermediate in the synthesis of natural isoflavones such as Tectorigenin, Irigenin, and Caviunin. It is classified as a phenol and polyphenol, and is an endogenous metabolite.
  • Degradation product of Iris Jorentina glucoside
  • Intermediate in natural isoflavone synthesis
  • Suitable for laboratory research applications
  • For research use only
Catalog No. 50-003-52408