Learn More
Carbosynth LLC. H-[Cys-Arg-Gly-Asp-Lys-Gly-Pro-Asp-Cys]-NH2, 1mg, A cell-penetrating peptide with tumor-specific binding., Disulfide bonds: Cys1-Cys9, MW: 947.07, Molecular formula: C35H58N14O13S2
SDP
Supplier:  Carbosynth LLC. RGD3761PI1MG
Encompass_Preferred
Synonyms: iRGD; cyclic CRGDKGPDC amide, H-[CRGDKGPDC]-NH2, Application: RGD Tumor Targeting Peptide, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: K Sugahara, et al., Science, 328, 1031 (2010)., Y. Ye, et al., Bioorg Med Chem Lett., 21, 1146 (2011)., T Kadonosono et al., J Control Release, 10(201), 14 (2015).
Catalog No. 50-168-6938