Learn More
Strem, An Ascensus Company CAS# 18827-81-1. 1g. Iridium carbonyl, min. 98%. MFCD00011064
SDP
Supplier:  Strem, An Ascensus Company 7718001G
Encompass
CAS# 18827-81-1. 1g. Iridium carbonyl, min. 98%. MFCD00011064. Molecular Weight: 1105.01. Molecular Formula: Ir4(CO)12. Color/form: yellow pwdr. Strem# 77-1800. http://www.strem.com/catalog/v/77-1800/
Catalog No. 50-901-13134