Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iridium(III) chloride hydrate | 14996-61-3 | MFCD00149749 | 98.0% | 298.58 g/mol (anhydrous basis) | IrCl3 · xH2O | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW021022100MG

Encompass_Preferred

Iridium(III) chloride hydrate (IrCl3 · xH2O, CAS 14996-61-3) is an inorganic/biochemical reagent used as a source of Ir(III) for synthesis and biological research. Supplied as a brown-to-black solid with a stated purity of 98%, it serves as a precursor for iridium complexes and is employed in coordination chemistry and select cancer research applications.

  • Source of Ir(III) for synthesis and catalyst preparation.
  • High purity (98%) for reproducible results.
  • Brown-to-black solid, easy to weigh and handle.
  • Formula IrCl3 · xH2O and molecular weight 298.58 g/mol (anhydrous basis).
  • Storage: store at 4°C, protect from light, store under nitrogen.
  • In solvent stability: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-01426


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.