Learn More
Medchemexpress LLC Iridium(III) chloride hydrate | 14996-61-3 | MFCD00149749 | 98.0% | 298.58 g/mol (anhydrous basis) | IrCl3 · xH2O | 250 MG

Supplier: Medchemexpress LLC 250MGHYW021022
Iridium(III) chloride hydrate (IrCl3 · xH2O) is a hydrated iridium trichloride biochemical reagent (CAS 14996-61-3) supplied as a solid with 98% purity for laboratory research. It is used in catalytic and biological studies, including reported applications in colon cancer research. Manufacturer guidance provides solubility and storage recommendations for proper handling.
- High purity (98%) reagent suitable for research applications.
- Hydrated solid form (IrCl3 · xH2O) for consistent handling.
- Soluble in water and DMSO with ultrasonic warming to improve dissolution.
- Available in small laboratory pack sizes for bench-scale work.
- Storage guidance provided: protect from light and store under inert atmosphere.
- Useful for catalytic and biological research, including colon cancer studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.