Learn More
Medchemexpress LLC Iriflophenone | 52591-10-3 | 5 MG

Supplier: Medchemexpress LLC HYN40105MG
Iriflophenone, isolated from Aquilaria sinensis, stimulates MCF-7 and T-47D human breast cancer cell proliferation. This product is for research use only and not sold to patients.
- Purity: 98.05%
- Molecular weight: 246.22
- Formula: C13H10O5
- Appearance: Solid
- Color: White to yellow
- Structure classification: Phenols, Polyphenols
- Initial source: Plants (Thymelaeaceae, Aquilaria sinensis (Lour.) Spreng.; Iridaceae, Belamcanda chinensis (Linn.) Redouté)
- Solubility (In Vitro): DMSO: 250 mg/mL (1015.35 mM; requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.