Learn More
Medchemexpress LLC Irinotecan-d10 hydrochloride | 718612-62-5 | 99.76% | 633.20 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY16562S11MG
Encompass_Preferred
Irinotecan-d10 (hydrochloride) is the deuterium labeled Irinotecan. Irinotecan ((+)-Irinotecan) is a topoisomerase I inhibitor, preventing religation of the DNA strand by binding to topoisomerase I-DNA complex. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Irinotecan.
  • Topoisomerase I inhibitor.
  • Prevents religation of the DNA strand by binding to topoisomerase I-DNA complex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
Catalog No. 50-003-93717