Learn More
Selleck Chemical LLC Irinotecan HCl Trihydrate 25mg 136572-09-3 CPT-11 HCl Trihydrate
SDP
Supplier:  Selleck Chemical LLC S221725MG
Encompass_Preferred
Irinotecan (CPT-11) prevents DNA from unwinding by inhibition of topoisomerase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2178