Learn More
TARGETMOL CHEMICALS INC Irinotecan hydrochloride trihy
SDP
Supplier:  TARGETMOL CHEMICALS INC T0486500MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Irinotecan hydrochloride trihydrate (CPT-11 HCl Trihydrate) keeps DNA from unwinding by inhibiting topoisomerase 1. Purity 99.52%
Catalog No. 50-288-0674