Learn More
Medchemexpress LLC HY-N0268 5mg Medchemexpress, Irisflorentin CAS:41743-73-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02685MG
Encompass_Preferred
Medchemexpress, HY-N0268 5mg Irisflorentin CAS:41743-73-1 Irisflorentin, a naturally occurring isoflavone, is an abundant active constituent in Rhizoma Belamcandae. Irisflorentin markedly reduces the transcriptional and translational levels of inducible nitric oxide synthase (iNOS) as well as the production of NO. Anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0245