Learn More
CPC Scientific Ac-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ENDT009A
Encompass
ONE-LETTER SEQUENCE: Ac-DKEAVYFAHLDIIWMOLECULAR FORMULA: C85H120N18O23MOLECULAR WEIGHT:1762STORAGE CONDITIONS: -20 5CRESEARCH AREA: CardiovascularREFERENCES: M. Kuwahara et al., Eur. J. Pharmacol., 296, 55 (1996)
Catalog No. 50-210-9244