Learn More
AdipoGen Iromycin A
SDP
Supplier:  AdipoGen BVT0262C500
Encompass_Preferred
Chemical. CAS 213137-53-2. Formula C19H29NO2. MW 303.4. Isolated from Streptomyces bottropensis Gö-Dra17. Pyridone metabolite. Nitric oxide synthase NOS inhibitor, showing selectivity for eNOS NOS III versus nNOS NOS I. Thaxtomine biosynthesis inhibitor. Mitochondrial electron transport chain inhibitor.
Catalog No. 50-256-2029