Learn More
Medchemexpress LLC Iron(5+), tetrakis(1-methylpyridiniumato) porphyrin pentachloride | 133314-07-5 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1203805MG
Encompass_Preferred
FeTMPyP is an iron-containing porphyrin complex used as a peroxynitrite (ONOO-) decomposition catalyst and scavenger in biochemical and pharmacological research. It reduces nitrative stress and has been reported to attenuate PARP over-activation and neuroinflammation in animal models.
  • Acts as a peroxynitrite decomposition catalyst and scavenger.
  • Useful for studying nitrative stress and related signaling pathways.
  • High purity for research applications.
  • Supplied as a 5 mg solid for small-scale experiments.
  • Stable when stored sealed at -20°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-34296