Learn More
Medchemexpress LLC Iron dextran | 9004-66-4 | 35.0-40.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY107928B5G
Encompass_Preferred
Iron dextran (Fe 35-40% w/w) is a colloidal ferric oxyhydroxide-dextran complex supplied as a biochemical reagent for laboratory research. It provides a concentrated source of elemental iron for assays and model studies and is intended for research use only, not for human therapeutic use.
  • Iron content 35-40% w/w.
  • Colloidal iron-dextran complex suitable for biochemical assays.
  • Supplied as a powder for laboratory preparation and formulation.
  • Available in laboratory pack sizes, including a 5 g option.
  • Safety and quality documents available: SDS and certificate of analysis.
  • For research use only; not for clinical use or sale to patients.
Catalog No. 50-002-42230