Learn More
Medchemexpress LLC Iron dextran | 9004-66-4 | 35.0-40.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY107928B500MG
Encompass_Preferred
Iron dextran (Fe 35-40% w/w) is a polymeric iron-dextran complex supplied as a biochemical reagent for research use. It is used in life-science studies such as investigations of iron supplementation and iron-deficiency anemia in animal models. The material is provided as a brown to reddish-brown solid and intended for laboratory research only.
  • Contains 35-40% elemental iron (w/w).
  • Polymeric iron-dextran complex suitable for biochemical studies.
  • Provided as a brown to reddish-brown solid.
  • Packaged as a 500 mg quantity.
  • Store dry at room temperature; in solution store at -80°C for up to 2 years or -20°C for up to 1 year.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-41785