Learn More
Medchemexpress LLC Iron dextran | 9004-66-4 | 35-40% | No defined chemical formula (polymeric iron-dextran complex) | 1 G
SDP
Supplier:  Medchemexpress LLC HY107928B1G
Encompass_Preferred
Iron dextran (Fe 35-40% w/w) is a polymeric iron-dextran complex supplied for laboratory research. It serves as an iron-containing biochemical reagent for assays and biological studies where a soluble iron source is required. Product documentation includes a safety data sheet and storage recommendations for research use only.
  • Iron content 35-40% w/w.
  • Polymeric iron-dextran complex with no single defined formula.
  • Intended for life-science research and biochemical assays.
  • Available in research pack sizes, including 1 g.
  • Safety data sheet and handling instructions provided.
  • Relative density approximately 1.17 g/cm3.
Catalog No. 50-002-42228