Learn More
Medchemexpress LLC Iron dextran | 9004-66-4 | MFCD00081553 | 35.0-40.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY107928B50MG
Encompass_Preferred
Iron dextran (Fe 35-40% w/w) is a dextran-bound iron complex supplied as a solid biochemical reagent for laboratory research. It delivers iron at approximately 35-40% w/w and is used as a reagent or biological material in life-science assays and preparative procedures. The material is polymeric, supplied in multiple pack sizes, and requires dry, protected storage per the SDS.
  • Provides iron at 35-40% w/w.
  • Polymeric ferric hydroxide-dextran complex.
  • Supplied as a solid suitable for assay and research use.
  • Available in small and bulk pack sizes for flexibility.
  • Recommended storage: keep sealed, cool, and protected from light.
  • For research use only; not for human therapeutic or diagnostic use.
Catalog No. 50-002-41723