Learn More
Medchemexpress LLC Iron dextran | 9004-66-4 | MFCD00081553 | ~5000 Da | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107928B100MG
Encompass_Preferred
Iron-Dextran (Fe 35-40% w/w) is a polymeric ferric hydroxide-dextran complex supplied as a solid reagent for life-science research. It provides an iron content of approximately 35-40% w/w and is commonly used in biochemical assays, iron-supplementation research, and studies of iron metabolism. The material is provided in multiple laboratory pack sizes and is accompanied by an SDS for safe handling.
Catalog No. 50-002-41722