Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iron(III) 2-methyl-4-oxo-4H-pyran-3-olate | 33725-54-1 | MFCD33402141 | 98.7% | 431.15 g/mol | C18H15FeO9 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY108017100MG

Encompass_Preferred

Ferric maltol is an orally active iron(III) complex used as a research reagent to study iron deficiency anemia, including cases associated with inflammatory bowel disease. It is supplied in solid and solution formats for laboratory use.

  • Orally active ferric iron (Fe3+) complex.
  • Used in research on iron deficiency anemia and inflammatory bowel disease.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • Molecular formula C18H15FeO9.
  • Molecular weight 431.15 g/mol.
  • CAS number 33725-54-1.

Catalog No. 50-002-42861


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.