Learn More
Medchemexpress LLC Iron(III) ferrocyanide | 14038-43-8 | MFCD00135663 | 99.0% | 859.23 | C18Fe7N18 | 10 G
SDP
Supplier:  Medchemexpress LLC HY106594A10G
Encompass_Preferred
Prussian blue insoluble (Iron(III) ferrocyanide) is an inorganic blue pigment and insoluble adsorbent used as an oral antidote for radiocesium and thallium poisoning. In research it functions as a contrast agent for photoacoustic and magnetic resonance imaging, shows antibacterial and anticancer activity, and exhibits photo-thermal and metabolic indicator properties in vitro.
  • High purity: 99.0%.
  • Molecular formula C18Fe7N18 and molar mass 859.23 g/mol.
  • Insoluble adsorbent effective for cesium and thallium decorporation.
  • Acts as a contrast agent for MRI and photoacoustic imaging.
  • Demonstrates photo-thermal effects and metabolic indicator behavior in vitro.
  • Stable solid with recommended refrigerated or frozen storage for extended shelf life.
Catalog No. 50-002-41030